Anti-ADAMTS5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: ADAMTS5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-128
- 77844-129
- Antigen Symbol:
- ADAMTS5
- ADAMTS5
- Antigen Synonyms:
- A disintegrin and metalloproteinase with thrombospondin motifs 11|A disintegrin and metalloproteinase with thrombospondin motifs 5|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 5|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase 2)|A disintegrin-like and metalloprotease with thrombospondin type 1 motif, 5|A Disintigrin And Metalloproteinase with ThromboSpondin motif-5|ADAM metallopeptidase with thrombospondin type 1 motif 5|ADAM TS 11|ADAM TS 5|ADAM TS5|ADAM-TS 11|ADAM-TS 5|ADAM-TS5|ADAMTS 11|ADAMTS 5|ADAMTS-11|ADAMTS-5|ADAMTS11|ADAMTS11, formerly|ADMP 2|ADMP-2|ADMP2|Aggrecanase 2|Aggrecanase-2|ATS5_HUMAN|FLJ36738|Implantin|ThromboSpondin motif-5
- A disintegrin and metalloproteinase with thrombospondin motifs 11|A disintegrin and metalloproteinase with thrombospondin motifs 5|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 5|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase 2)|A disintegrin-like and metalloprotease with thrombospondin type 1 motif, 5|A Disintigrin And Metalloproteinase with ThromboSpondin motif-5|ADAM metallopeptidase with thrombospondin type 1 motif 5|ADAM TS 11|ADAM TS 5|ADAM TS5|ADAM-TS 11|ADAM-TS 5|ADAM-TS5|ADAMTS 11|ADAMTS 5|ADAMTS-11|ADAMTS-5|ADAMTS11|ADAMTS11, formerly|ADMP 2|ADMP-2|ADMP2|Aggrecanase 2|Aggrecanase-2|ATS5_HUMAN|FLJ36738|Implantin|ThromboSpondin motif-5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UNA0
- Q9UNA0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human A disintegrin and metalloproteinase with thrombospondin motifs 5 protein (535-646AA)
- Recombinant Human A disintegrin and metalloproteinase with thrombospondin motifs 5 protein (535-646AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice