Anti-PDZRN3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDZRN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-548
- 77844-547
- Antigen Symbol:
- PDZRN3
- PDZRN3
- Antigen Synonyms:
- E3 ubiquitin-protein ligase KIAA1095|Ligand of Numb protein X 3|likely ortholog of mouse semaF cytoplasmic domain associated protein 3|LNX3|PDZ domain containing ring finger 3|PDZ domain-containing RING finger protein 3|Protein SEMACAP3|PZRN3_HUMAN|SEMACAP3|Semaphorin cytoplasmic domain associated protein|Semaphorin cytoplasmic domain-associated protein 3|SEMCAP3
- E3 ubiquitin-protein ligase KIAA1095|Ligand of Numb protein X 3|likely ortholog of mouse semaF cytoplasmic domain associated protein 3|LNX3|PDZ domain containing ring finger 3|PDZ domain-containing RING finger protein 3|Protein SEMACAP3|PZRN3_HUMAN|SEMACAP3|Semaphorin cytoplasmic domain associated protein|Semaphorin cytoplasmic domain-associated protein 3|SEMCAP3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UPQ7
- Q9UPQ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human E3 ubiquitin-protein ligase PDZRN3 protein (133-404AA)
- Recombinant Human E3 ubiquitin-protein ligase PDZRN3 protein (133-404AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice