Anti-BMPR1A Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77851-916
New Product
- Antigen Symbol:BMPR1A
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P36894
- Antigen Synonyms:10q23del|Activin A receptor type II like kinase 3|Activin receptor like kinase 3|Activin receptor-like kinase 3|ACVRLK 3|ACVRLK3|ALK 3|ALK-3|ALK3|BMP type-1A receptor|BMPR 1A|Bmpr|BMPR-1A|BMR1A_HUMAN|Bone morphogenetic protein receptor type IA|Bone morphogenetic protein receptor type IA precursor|Bone morphogenetic protein receptor type-1A|BR 1a|BR1a|CD 292|CD292|CD292 antigen|EC 2.7.11.30|Serine threonine protein kinase receptor R5|Serine threonine protein kinase receptor R5 precursor|Serine/threonine-protein kinase receptor R5|SKR 5|SKR5|zBMPR IA|zBMPRIA
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Bone morphogenetic protein receptor type-1A protein (177-532AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: BMPR1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human