Anti-USH1C Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: USH1C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-815
- 77845-816
- Antigen Symbol:
- USH1C
- USH1C
- Antigen Synonyms:
- AIE 75|AIE75|Antigen NY CO 38/NY CO 37|Antigen NY-CO-38/NY-CO-37|Autoimmune enteropathy related antigen AIE 75|Autoimmune enteropathy related antigen AIE75|Autoimmune enteropathy-related antigen AIE-75|Deafness autosomal recessive 18|DFNB 18|DFNB18|Harmonin|NY CO 37|NY CO 38|PDZ 45|PDZ 73|PDZ 73 protein|PDZ 73/NY CO 38|PDZ45|PDZ73|PDZ73 protein|Protein PDZ-73|Renal carcinoma antigen NY REN 3|Renal carcinoma antigen NY-REN-3|USH 1C|USH1C_HUMAN|Ush1cpst|Usher syndrome 1C (autosomal recessive severe)|Usher syndrome 1C|Usher syndrome type 1C protein|Usher syndrome type-1C protein
- AIE 75|AIE75|Antigen NY CO 38/NY CO 37|Antigen NY-CO-38/NY-CO-37|Autoimmune enteropathy related antigen AIE 75|Autoimmune enteropathy related antigen AIE75|Autoimmune enteropathy-related antigen AIE-75|Deafness autosomal recessive 18|DFNB 18|DFNB18|Harmonin|NY CO 37|NY CO 38|PDZ 45|PDZ 73|PDZ 73 protein|PDZ 73/NY CO 38|PDZ45|PDZ73|PDZ73 protein|Protein PDZ-73|Renal carcinoma antigen NY REN 3|Renal carcinoma antigen NY-REN-3|USH 1C|USH1C_HUMAN|Ush1cpst|Usher syndrome 1C (autosomal recessive severe)|Usher syndrome 1C|Usher syndrome type 1C protein|Usher syndrome type-1C protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Harmonin protein (1-533AA)
- Recombinant Human Harmonin protein (1-533AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice