Anti-CCRL2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77852-583
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CCRL2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O00421
- Antigen Synonyms:C-C chemokine receptor-like 2|CCRL2_HUMAN|Chemokine (C-C motif) receptor like 2|Chemokine receptor CCR11|Chemokine receptor X|CKRX|CRAM A|CRAM B|CRAMA|CRAMB|FLJ55815|HCR|MGC116710|MGC34104|Putative MCP 1 chemokine receptor|Putative MCP-1 chemokine receptor
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human C-C chemokine receptor-like 2 protein (1-43AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CCRL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human