Anti-cyp26a1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77853-903
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:cyp26a1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Zebrafish
- Size:50 µg
- Form:Liquid
- Gene ID:P79739
- Antigen Synonyms:cyp26Cytochrome P450 26A1|EC 1.14.13.-|Cytochrome P450RAI|Retinoic acid 4-hydroxylase|Retinoic acid-metabolizing cytochrome
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Zebrafish Cytochrome P450 26A1 protein (244-371AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: cyp26a1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish