Anti-PDSS2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: PDSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77847-682
- 77847-681
- Antigen Symbol:
- PDSS2
- PDSS2
- Antigen Synonyms:
- All-trans-decaprenyl-diphosphate synthase subunit 2|bA59I9.3|C6orf210|Candidate tumor suppressor protein|chromosome 6 open reading frame 210|Decaprenyl pyrophosphate synthase subunit 2|decaprenyl pyrophosphate synthetase subunit 2|Decaprenyl-diphosphate synthase subunit 2|DLP1|DLP1_HUMAN|hDLP1|Pdss2|prenyl (decaprenyl) diphosphate synthase|subunit 2|subunit 2 of decaprenyl diphosphate synthase
- All-trans-decaprenyl-diphosphate synthase subunit 2|bA59I9.3|C6orf210|Candidate tumor suppressor protein|chromosome 6 open reading frame 210|Decaprenyl pyrophosphate synthase subunit 2|decaprenyl pyrophosphate synthetase subunit 2|Decaprenyl-diphosphate synthase subunit 2|DLP1|DLP1_HUMAN|hDLP1|Pdss2|prenyl (decaprenyl) diphosphate synthase|subunit 2|subunit 2 of decaprenyl diphosphate synthase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86YH6
- Q86YH6
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human PDSS2
- Synthetic peptide of Human PDSS2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice