SARS-CoV-2 Recombinant SARS-CoV-2 Spike Glycoprotein (from E. coli)
Supplier: Antibodies.com
Certificates
About this item
Partial length SARS-CoV-2 SARS-CoV-2 Spike Glycoprotein, expressed in E. coli, suitable for SDS-PAGE, ELISA, Immunogen Grade.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Storage Conditions:Upon delivery aliquot and store at -20 °C. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P0DTC2
- Carrier-Free:N
- Protein/Peptide Name:SARS-CoV-2 Spike Glycoprotein
- Molecular Weight:32 kDa (by SDS-PAGE)
- Sequence:MHHHHHHSSGLVPRGSGMKETAAAKFERQHMDSPDLGTDDDDKAMADIGSEFVEKGIYQTSNFRVQPTESIVRFPNITNLCPFGEVFNATRFASVYAWNRKRISNCVADYSVLYNSASFSTFKCYGVSPTKLNDLCFTNVYADSFVIRGDEVRQIAPGQTGKIADYNYKLPDDFTGCVIAWNSNNLDSKVGGNYNYLYRLFRKSNLKPFERDISTEIYQAGSTPCNGVEGFNCYFPLQSYGFQPTNGVGYQPYRVVVLSFELLHAPATVCGPKKSTNLVKNKCVNF
- Concentration:1 mg/ml
- Formulation:Supplied in 10 mM phosphate buffer, pH 7.5, with 6 M urea
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:SDS-PAGE, ELISA, Immunogen Grade
Specifications
Frequently Bought Together