Anti-STRAP Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: STRAP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77845-661
- 77845-662
- Antigen Symbol:
- STRAP
- STRAP
- Antigen Synonyms:
- MAP activator with WD repeats|MAWD|PTWD|Serine-threonine kinase receptor-associated protein|serine/threonine kinase receptor associated protein|STRAP_HUMAN|UNR-interacting protein|UNRIP|WD 40 repeat protein PT WD|WD-40 repeat protein PT-WD
- MAP activator with WD repeats|MAWD|PTWD|Serine-threonine kinase receptor-associated protein|serine/threonine kinase receptor associated protein|STRAP_HUMAN|UNR-interacting protein|UNRIP|WD 40 repeat protein PT WD|WD-40 repeat protein PT-WD
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3F4
- Q9Y3F4
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine-threonine kinase receptor-associated protein (1-350AA)
- Recombinant Human Serine-threonine kinase receptor-associated protein (1-350AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together