Anti-UFC1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UFC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-361
- 77846-362
- Antigen Symbol:
- UFC1
- UFC1
- Antigen Synonyms:
- CGI-126|HSPC155|Ubiquitin fold modifier conjugating enzyme 1|Ubiquitin-fold modifier-conjugating enzyme 1|UFC1|UFC1_HUMAN|Ufm1 conjugating enzyme 1|Ufm1-conjugating enzyme 1
- CGI-126|HSPC155|Ubiquitin fold modifier conjugating enzyme 1|Ubiquitin-fold modifier-conjugating enzyme 1|UFC1|UFC1_HUMAN|Ufm1 conjugating enzyme 1|Ufm1-conjugating enzyme 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3C8
- Q9Y3C8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin-fold modifier-conjugating enzyme 1 protein (1-167AA)
- Recombinant Human Ubiquitin-fold modifier-conjugating enzyme 1 protein (1-167AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice