Anti-USP22 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: USP22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-700
- 77844-699
- Antigen Symbol:
- USP22
- USP22
- Antigen Synonyms:
- Deubiquitinating enzyme 22|KIAA1063|Ubiquitin carboxyl terminal hydrolase 22|Ubiquitin carboxyl-terminal hydrolase 22|Ubiquitin specific peptidase 22|Ubiquitin specific peptidase 3 like|Ubiquitin specific processing protease 22|Ubiquitin specific protease 22|Ubiquitin thioesterase 22|Ubiquitin thiolesterase 22|Ubiquitin-specific-processing protease 22|UBP22_HUMAN|USP 22|Usp22|USP3L
- Deubiquitinating enzyme 22|KIAA1063|Ubiquitin carboxyl terminal hydrolase 22|Ubiquitin carboxyl-terminal hydrolase 22|Ubiquitin specific peptidase 22|Ubiquitin specific peptidase 3 like|Ubiquitin specific processing protease 22|Ubiquitin specific protease 22|Ubiquitin thioesterase 22|Ubiquitin thiolesterase 22|Ubiquitin-specific-processing protease 22|UBP22_HUMAN|USP 22|Usp22|USP3L
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UPT9
- Q9UPT9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin carboxyl-terminal hydrolase 22 protein (330-478AA)
- Recombinant Human Ubiquitin carboxyl-terminal hydrolase 22 protein (330-478AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice