Anti-CGN Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77853-029
New Product
- Antigen Symbol:CGN
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9P2M7
- Antigen Synonyms:6330408J11Rik|AI647528|AI987749|CING_HUMAN|Cingulin|DKFZp779N1112|FLJ39281|KIAA1319|MGC118157
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cingulin protein (92-252AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: CGN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human