Anti-NDUFA12 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:10000, IHC: 1:50 to 1:200
Type:
Antigen: NDUFA12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-604
- 77847-603
- Antigen Symbol:
- NDUFA12
- NDUFA12
- Antigen Synonyms:
- 13 kDa differentiation associated protein|13 kDa differentiation-associated protein|2410011G03Rik|AW112974|B17.2|CI-B17.2|CIB17.2|Complex I B17.2|Complex I B17.2 subunit|Complex I-B17.2|DAP13|MGC107642|MGC7999|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 12|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12|NADH ubiquinone oxidoreductase subunit B17.2|NADH-ubiquinone oxidoreductase subunit B17.2|NDUAC_HUMAN|RGD1311462
- 13 kDa differentiation associated protein|13 kDa differentiation-associated protein|2410011G03Rik|AW112974|B17.2|CI-B17.2|CIB17.2|Complex I B17.2|Complex I B17.2 subunit|Complex I-B17.2|DAP13|MGC107642|MGC7999|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 12|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12|NADH ubiquinone oxidoreductase subunit B17.2|NADH-ubiquinone oxidoreductase subunit B17.2|NDUAC_HUMAN|RGD1311462
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UI09
- Q9UI09
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human NDUFA12
- Synthetic peptide of Human NDUFA12
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice