Anti-ACSL5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:500 to 1:1000
Type:
Antigen: ACSL5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Specifications
- Catalog No:
- 77844-465
- 77844-464
- Antigen Symbol:
- ACSL5
- ACSL5
- Antigen Synonyms:
- ACS5|FACL5|UNQ633/PRO1250|Long-chain-fatty-acid--CoA ligase 5|EC 6.2.1.3|Arachidonate--CoA ligase|EC 6.2.1.15|Long-chain acyl-CoA synthetase 5|LACS 5
- ACS5|FACL5|UNQ633/PRO1250|Long-chain-fatty-acid--CoA ligase 5|EC 6.2.1.3|Arachidonate--CoA ligase|EC 6.2.1.15|Long-chain acyl-CoA synthetase 5|LACS 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULC5
- Q9ULC5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Long-chain-fatty-acid--CoA ligase 5 protein (33-683AA)
- Recombinant human Long-chain-fatty-acid--CoA ligase 5 protein (33-683AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice