Anti-NLK Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: NLK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-742
- 77843-741
- Antigen Symbol:
- NLK
- NLK
- Antigen Synonyms:
- DKFZp761G1211|FLJ21033|LAK1|Nemo like kinase|Nemo-like kinase|Nlk|NLK_HUMAN|Protein LAK1|Serine/threonine protein kinase NLK|Serine/threonine-protein kinase NLK
- DKFZp761G1211|FLJ21033|LAK1|Nemo like kinase|Nemo-like kinase|Nlk|NLK_HUMAN|Protein LAK1|Serine/threonine protein kinase NLK|Serine/threonine-protein kinase NLK
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UBE8
- Q9UBE8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase NLK protein (278-527AA)
- Recombinant Human Serine/threonine-protein kinase NLK protein (278-527AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice