Anti-ERGIC3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77845-557
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ERGIC3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y282
- Antigen Synonyms:2310015B14Rik|AV318804|C20orf47|CGI 54|D2Ucla1|dJ477O4.2|DKFZp547A2190|Endoplasmic reticulum Golgi intermediate compartment protein 3|endoplasmic reticulum localized protein ERp43|Endoplasmic reticulum-Golgi intermediate compartment protein 3|ERGI3_HUMAN|ERGIC and golgi 3|ERV46|NY BR 84|PRO0989|RP23-220D12.2|RP3-477O4.1|SDBCAG84|Serologically defined breast cancer antigen 84|Serologically defined breast cancer antigen NY BR 84|Serologically defined breast cancer antigen NY-BR-84
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Endoplasmic reticulum-Golgi intermediate compartment protein 3 protein (47-341AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ERGIC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human