Anti-SSX2IP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: SSX2IP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-471
- 77846-472
- Antigen Symbol:
- SSX2IP
- SSX2IP
- Antigen Synonyms:
- ADIP|ADIP_HUMAN|Afadin and alpha actinin binding protein|Afadin DIL domain interacting protein|Afadin DIL domain-interacting protein|Afadin- and alpha-actinin-binding protein|FLJ10848|KIAA0923|MGC75026|OTTHUMP00000011574|SSX 2 interacting protein|SSX2 interacting protein|SSX2-interacting protein|SSX2IP|Synovial sarcoma X breakpoint 2 interacting protein
- ADIP|ADIP_HUMAN|Afadin and alpha actinin binding protein|Afadin DIL domain interacting protein|Afadin DIL domain-interacting protein|Afadin- and alpha-actinin-binding protein|FLJ10848|KIAA0923|MGC75026|OTTHUMP00000011574|SSX 2 interacting protein|SSX2 interacting protein|SSX2-interacting protein|SSX2IP|Synovial sarcoma X breakpoint 2 interacting protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2D8
- Q9Y2D8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Afadin- and alpha-actinin-binding protein (485-614AA)
- Recombinant Human Afadin- and alpha-actinin-binding protein (485-614AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice