Anti-GSK3B (phospho-Ser9) HEK293F cells Recombinant Monoclonal Antibody [clone: 3A6]
77850-230
New Product
- Antigen Symbol:GSK3B
- Clonality:Recombinant Monoclonal
- Clone:3A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P49841
- Antigen Synonyms:Glycogen synthase kinase-3 beta|GSK-3 beta|Serine/threonine-protein kinase GSK3B
- Modification:phospho-Ser9
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human Phospho-GSK3B (Ser9)
- Purification:Affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:50 to 1:200, IF: 1:20 to 1:200
Type:
Antigen: GSK3B
Clonality: Recombinant Monoclonal
Clone: 3A6
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human