Anti-S100A4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: S100A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77847-802
- 77847-801
- Antigen Symbol:
- S100A4
- S100A4
- Antigen Synonyms:
- 18A2|42A|calcium Placental protein|Calvasculin|CAPL|Fibroblast specific protein 1 (FSP1)|Fibroblast specific protein 1|Fibroblast specific protein|FSP1|Leukemia multidrug resistance associated protein|Malignant transformation suppression 1 (MTS1)|Malignant transformation suppression 1|Metastasin|MTS1|OTTHUMP00000015467|OTTHUMP00000015468|P9KA|PEL98|Placental calcium-binding protein|Protein Mts1|Protein S100 A4|Protein S100-A4|S100 calcium binding protein A4 (calcium protein|murine placental homolog)|S100 calcium binding protein A4|S100 calcium-binding protein A4|S10A4_HUMAN
- 18A2|42A|calcium Placental protein|Calvasculin|CAPL|Fibroblast specific protein 1 (FSP1)|Fibroblast specific protein 1|Fibroblast specific protein|FSP1|Leukemia multidrug resistance associated protein|Malignant transformation suppression 1 (MTS1)|Malignant transformation suppression 1|Metastasin|MTS1|OTTHUMP00000015467|OTTHUMP00000015468|P9KA|PEL98|Placental calcium-binding protein|Protein Mts1|Protein S100 A4|Protein S100-A4|S100 calcium binding protein A4 (calcium protein|murine placental homolog)|S100 calcium binding protein A4|S100 calcium-binding protein A4|S10A4_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P26447
- P26447
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human S100A4
- Synthetic peptide of Human S100A4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice