Anti-CXCR3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:1000 to 1:5000, IHC:1:25 to 1:100
Type:
Antigen: CXCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77849-781
- 77849-782
- Antigen Symbol:
- CXCR3
- CXCR3
- Antigen Synonyms:
- C-X-C chemokine receptor type 3|CD 183|CD182|CD183|Chemokine (C X C motif) receptor 3|Chemokine (C X C) receptor 3|Chemokine CXC Motif Receptor 3|CKR L2|CKR-L2|CKRL2|CMKAR3|CXC-R3|CXCR-3|CXCR3|CXCR3_HUMAN|G Protein Coupled Receptor 9 |G protein-coupled receptor 9|GPR9|Interferon-inducible protein 10 receptor|IP-10 receptor|IP10|IP10 R|IP10 receptor|IP10-R|IP10R|Mig R|Mig receptor|Mig-R|MIGR
- C-X-C chemokine receptor type 3|CD 183|CD182|CD183|Chemokine (C X C motif) receptor 3|Chemokine (C X C) receptor 3|Chemokine CXC Motif Receptor 3|CKR L2|CKR-L2|CKRL2|CMKAR3|CXC-R3|CXCR-3|CXCR3|CXCR3_HUMAN|G Protein Coupled Receptor 9 |G protein-coupled receptor 9|GPR9|Interferon-inducible protein 10 receptor|IP-10 receptor|IP10|IP10 R|IP10 receptor|IP10-R|IP10R|Mig R|Mig receptor|Mig-R|MIGR
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49682
- P49682
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CXCR3
- Fusion protein of Human CXCR3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice