Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-C9ORF72/C9RANT Mouse Monoclonal Antibody [clone: 5E9]
Anti-C9ORF72/C9RANT Mouse Monoclonal Antibody [clone: 5E9]
Catalog # 103258-500
Supplier:  MilliporeSigma
Anti-C9ORF72/C9RANT Mouse Monoclonal Antibody [clone: 5E9]
Catalog # 103258-500
Supplier:  MilliporeSigma
Supplier Number:  MABN889

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    C9ORF72/C9RANT (poly-GA)
  • Clonality:
    Monoclonal
  • Clone:
    5E9
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Multiple,
    Human
  • Size:
    50 µl
  • Gene ID:
    Q96LT7
  • Shipping Temperature:
    Wet ice

Specifications

About this item

Expansion of a GGGGCC hexanucleotide repeat sequence in the non-coding region of human chromosome 9 open reading frame 72 or C9orf72 (also known as ALSFTD, FTDALS; Gene ID 203228) is the most common genetic abnormality in familial and sporadic frontotemporal dementia (FTD) and motor neuron disease (MND), with amyotrophic lateral sclerosis (ALS) as the most frequent form. The number of hexanucleotide repeats in the normal population ranges from 2 to 24, whereas up to several thousand repeats (700 to 4,400 repeats) are found in the pathologically expanded allele.

Unconventional repeat-associated non-ATG-initiated translation (RANT) of the GGGGCC repeats in the three alternate reading frames generates three polypeptides, each composed of repeating units of two amino acids (dipeptide repeats, DPRs), glycine-alanine (GA), glycine-proline (GP), and glycine-arginine (GR), respectively. These DPR proteins form inclusions particularly in the cerebellar cortex, hippocampus and cerebral neocortex. These inclusions are also immunoreactive for markers of the ubiquitin proteasome system (UPS), including ubiquitin, ubiquilins, and p62, but is distinct from inclusions containing TDP-43 that is also found in FTLD-TDP (frontotemporal lobar degeneration with TDP-43 pathology) and ALS cases. Antibodies against poly-GA, poly-GP, and poly-GR sequence are useful for characterizing the neuroanatomical distribution and clinico-pathological association of DPR pathology with C9ORF72 mutations.

Type:
Antigen: C9ORF72/C9RANT (poly-GA)
Clonality: Monoclonal
Clone: 5E9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Multiple, Human