Anti-MAGEB4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:15 to 1:50
Type:
Antigen: MAGEB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77847-037
- 77847-036
- Antigen Symbol:
- MAGEB4
- MAGEB4
- Antigen Synonyms:
- cancer/testis antigen family 3, member 6|CT3.6|MAGB4_HUMAN|MAGE B4 antigen|MAGE-B4 antigen|MAGEB4|melanoma antigen family B, 4|Melanoma associated antigen B4|Melanoma-associated antigen B4|MGC33144
- cancer/testis antigen family 3, member 6|CT3.6|MAGB4_HUMAN|MAGE B4 antigen|MAGE-B4 antigen|MAGEB4|melanoma antigen family B, 4|Melanoma associated antigen B4|Melanoma-associated antigen B4|MGC33144
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15481
- O15481
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human MAGEB4
- Fusion protein of Human MAGEB4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice