Anti-DENND2D Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DENND2D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-185
- 77843-184
- Antigen Symbol:
- DENND2D
- DENND2D
- Antigen Synonyms:
- 2010308M01Rik|AI451421|DEN2D_HUMAN|DENN domain containing 2D|DENN domain-containing protein 2D|DENN/MADD domain containing 2D|FLJ22457|MGC6915|RGD1305120|RP5-1180E21.2
- 2010308M01Rik|AI451421|DEN2D_HUMAN|DENN domain containing 2D|DENN domain-containing protein 2D|DENN/MADD domain containing 2D|FLJ22457|MGC6915|RGD1305120|RP5-1180E21.2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H6A0
- Q9H6A0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DENN domain-containing protein 2D protein (1-233AA)
- Recombinant Human DENN domain-containing protein 2D protein (1-233AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice