Anti-SLC5A11 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: SLC5A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77848-944
- 77848-943
- Antigen Symbol:
- SLC5A11
- SLC5A11
- Antigen Synonyms:
- homolog of rabbit KST1|KST1|Na(+)/myo-inositol cotransporter 2|na+/myo-inositol cotransporter 2|putative sodium-coupled cotransporter RKST1|RKST1|SC5AB_HUMAN|SGLT6|SMIT2|Sodium-dependent glucose cotransporter|Sodium/glucose cotransporter KST1|Sodium/myo-inositol cotransporter 2|Sodium/myo-inositol transporter 2|solute carrier family 5 (sodium/glucose cotransporter)|member 11|Solute carrier family 5 member 11
- homolog of rabbit KST1|KST1|Na(+)/myo-inositol cotransporter 2|na+/myo-inositol cotransporter 2|putative sodium-coupled cotransporter RKST1|RKST1|SC5AB_HUMAN|SGLT6|SMIT2|Sodium-dependent glucose cotransporter|Sodium/glucose cotransporter KST1|Sodium/myo-inositol cotransporter 2|Sodium/myo-inositol transporter 2|solute carrier family 5 (sodium/glucose cotransporter)|member 11|Solute carrier family 5 member 11
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WWX8
- Q8WWX8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SLC5A11
- Synthetic peptide of Human SLC5A11
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice