Anti-ASGR2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77851-470
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ASGR2
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P07307
- Antigen Synonyms:ASGP R| ASGP-R 2| ASGPR 2| ASGPR2| Asgr2| ASGR2_HUMAN| Asialoglycoprotein receptor 2| C type lectin domain family 4 member H2| C-type lectin domain family 4 member H2| CLEC4H2| HBxAg-binding protein| HBXBP| Hepatic lectin 2| Hepatic lectin H2| Hepatic lectin R2/3| HL-2| Hs.1259| L H2| LH2| MGC108546| MHL 2| RHL2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Asialoglycoprotein receptor 2 protein (63-286AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: ASGR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human