Anti-APOBEC3G Rabbit Recombinant Polyclonal Antibody
77851-298
New Product
- Antigen Symbol:APOBEC3G
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9HC16
- Antigen Synonyms:A3G|ABC3G_HUMAN|APOBEC related cytidine deaminase|APOBEC related protein|APOBEC-related cytidine deaminase|APOBEC-related protein 9|APOBEC-related protein|Apolipoprotein B editing enzyme catalytic polypeptide like 3G|Apolipoprotein B mRNA editing enzyme catalytic polypeptide 3G|Apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3G|Apolipoprotein B mRNA editing enzyme catalytic subunit 3G|apolipoprotein B mRNA editing enzyme cytidine deaminase|apolipoprotein B mRNA editing enzyme|catalytic polypeptide-like|ARCD|ARP-9|ARP9|bK150C2.7|CEM-15|CEM15|deoxycytidine deaminase|dJ494G10.1|DNA dC dU editing enzyme APOBEC 3G|DNA dC->dU editing enzyme|DNA dC->dU-editing enzyme APOBEC-3G|EC 3.5.4.|FLJ12740|MDS019|OTTHUMP00000028911|phorbolin-like protein|phorbolin-like protein MDS019
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human DNA dC->dU-editing enzyme APOBEC-3G protein (1-384AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: APOBEC3G
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human