Anti-BANF1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77851-787
New Product
- Antigen Symbol:BANF1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75531
- Antigen Synonyms:BAF|BAF_HUMAN|BANF 1|Barrier to autointegration factor 1|Barrier to autointegration factor|Barrier-to-autointegration factor|BCRG 1|BCRG1|BCRP 1|BCRP1|Breakpoint cluster region protein 1|D14S1460|MGC111161|NGPS
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Barrier-to-autointegration factor protein (2-89AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
0
Type:
Antigen: BANF1
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human