Anti-ALDH6A1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77851-053
New Product
- Antigen Symbol:ALDH6A1
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q02252
- Antigen Synonyms:1110038I05Rik|AI314632|Aldehyde dehydrogenase 6 family member A1|Aldehyde dehydrogenase family 6 member A1|Aldehyde dehydrogenase family 6, subfamily A1|Malonate semialdehyde dehydrogenase|Malonate-semialdehyde dehydrogenase [acylating]|Methylmalonate semialdehyde dehydrogenase [acylating] mitochondrial|Methylmalonate-semialdehyde dehydrogenase [acylating]|MGC37325|MGC40271|Mitochondrial acylating methylmalonate semialdehyde dehydrogenase|mitochondrial|MMSA_HUMAN|MMSADHA|MMSDH
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial protein (274-399AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Type:
Antigen: ALDH6A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human