Anti-ZFP36L2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: ZFP36L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77849-756
- 77849-755
- Antigen Symbol:
- ZFP36L2
- ZFP36L2
- Antigen Synonyms:
- BRF2|Butyrate response factor 2|C3H1 type-like 2|EGF response factor 2|EGF-response factor 2|ERF 2|ERF-2|ERF2|Protein TIS11D|RNF162C|TIS11D|TISD_HUMAN|ZFP36 like 2|ZFP36 ring finger protein like 2|ZFP36-like 2|Zinc finger protein 36|Zinc finger protein 36, C3H type like 1|zinc finger protein 36, C3H type like 2|Zinc finger protein 36, C3H1 type like 2|Zinc finger protein, C3H type, 36 like 2
- BRF2|Butyrate response factor 2|C3H1 type-like 2|EGF response factor 2|EGF-response factor 2|ERF 2|ERF-2|ERF2|Protein TIS11D|RNF162C|TIS11D|TISD_HUMAN|ZFP36 like 2|ZFP36 ring finger protein like 2|ZFP36-like 2|Zinc finger protein 36|Zinc finger protein 36, C3H type like 1|zinc finger protein 36, C3H type like 2|Zinc finger protein 36, C3H1 type like 2|Zinc finger protein, C3H type, 36 like 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P47974
- P47974
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ZFP36L2
- Synthetic peptide of Human ZFP36L2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice