Anti-ITGAE Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: ITGAE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-153
- 77848-152
- Antigen Symbol:
- ITGAE
- ITGAE
- Antigen Synonyms:
- A530055J10|alpha-E1|alpha-M290|aM290|Antigen CD103|CD 103|CD103|CD103 antigen|HML 1 antigen|HML-1 antigen|HML1 antigen|Human mucosal lymphocyte antigen 1|alpha subunit|Huminae|Integrin alpha E|Integrin alpha E heavy chain|Integrin alpha E light chain|epithelial-associated|Integrin alpha E1|Integrin alpha IEL|Integrin alpha M290|Integrin alpha-E heavy chain|Integrin alpha-IEL|Integrin|alpha E (antigen CD103|alpha polypeptide)|alpha E|ITAE_HUMAN|MGC141996|MGC183601|Mucosal lymphocyte 1 antigen|Mucosal lymphocyte antigen 1
- A530055J10|alpha-E1|alpha-M290|aM290|Antigen CD103|CD 103|CD103|CD103 antigen|HML 1 antigen|HML-1 antigen|HML1 antigen|Human mucosal lymphocyte antigen 1|alpha subunit|Huminae|Integrin alpha E|Integrin alpha E heavy chain|Integrin alpha E light chain|epithelial-associated|Integrin alpha E1|Integrin alpha IEL|Integrin alpha M290|Integrin alpha-E heavy chain|Integrin alpha-IEL|Integrin|alpha E (antigen CD103|alpha polypeptide)|alpha E|ITAE_HUMAN|MGC141996|MGC183601|Mucosal lymphocyte 1 antigen|Mucosal lymphocyte antigen 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P38570
- P38570
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ITGAE
- Fusion protein of Human ITGAE
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice