Anti-ATP6V1B1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-641
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP6V1B1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Form:Liquid
- Gene ID:P15313
- Antigen Synonyms:Atp6b1|ATPase|H+ transporting|lysosomal (vacuolar proton pump)|beta 56/58 kDa|isoform 1|lysosomal 56/58kDa|V1 subunit B|Endomembrane proton pump 58 kDa subunit|H(+) transporting two sector ATPase 58kD subunit|H+ ATPase beta 1 subunit|kidney isoform|MGC32642|RTA1B|V ATPase B1 subunit|V type proton ATPase subunit B kidney isoform|V type proton ATPase subunit B|V-ATPase subunit B 1|V-type proton ATPase subunit B|Vacuolar proton pump 3|Vacuolar proton pump subunit 3|Vacuolar proton pump subunit B 1|VATB|VATB1_HUMAN|VMA2|Vpp 3|Vpp3
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human V-type proton ATPase subunit B, kidney isoform protein (1-280AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: ATP6V1B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse