Anti-SERPINA9 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: SERPINA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-279
- 77845-280
- Antigen Symbol:
- SERPINA9
- SERPINA9
- Antigen Synonyms:
- Centerin|GCET1|Germinal center B cell expressed transcript 1|Germinal center B-cell-expressed transcript 1 protein|Seprin A9|Serine proteinase inhibitor A11|Serpin A9|Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 9|Serpin peptidase inhibitor clade A member 9|SERPINA11|SERPINA11b|SPA9_HUMAN
- Centerin|GCET1|Germinal center B cell expressed transcript 1|Germinal center B-cell-expressed transcript 1 protein|Seprin A9|Serine proteinase inhibitor A11|Serpin A9|Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 9|Serpin peptidase inhibitor clade A member 9|SERPINA11|SERPINA11b|SPA9_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86WD7
- Q86WD7
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SERPINA9
- Synthetic peptide of Human SERPINA9
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice