Anti-CA14 Rabbit Recombinant Polyclonal Antibody
Catalog # 77844-498
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CA14
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9ULX7
- Antigen Synonyms:CA 14|CA XIV|CA-XIV|CAH14_HUMAN|Carbonate dehydratase XIV|Carbonic anhydrase 14|Carbonic anhydrase XIV|Carbonic dehydratase|CAXIV|UNQ690/PRO1335
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Carbonic anhydrase 14 protein (16-290AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: CA14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse