Anti-APOA1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77851-272
New Product
- Antigen Symbol:APOA1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Form:Liquid
- Gene ID:P15497
- Antigen Synonyms:APOA1Apolipoprotein A-I|Apo-AI|ApoA-I|Apolipoprotein A1) [Cleaved into: Proapolipoprotein A-I|ProapoA-I)|Truncated apolipoprotein A-I]
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Bovine Apolipoprotein A-I protein (25-265AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Type:
Antigen: APOA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine