Anti-CORO1A Rabbit Recombinant Polyclonal Antibody (Biotin)
Catalog # 77853-431
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CORO1A
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P31146
- Antigen Synonyms:Actin binding protein|CLABP|Clipin A|Clipin-A|CLIPINA|COR1A_HUMAN|Coronin 1|Coronin actin binding protein 1A|Coronin like protein A|Coronin like protein p57|Coronin-1A|Coronin-like protein A|Coronin-like protein p57|FLJ41407|HCORO1|MGC117380|OTTHUMP00000163017|p57|TACO|Tryptophan aspartate containing coat protein|Tryptophan aspartate-containing coat protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Coronin-1A protein (363-432AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CORO1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human