Anti-SF3B6 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SF3B6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-533
- 77846-534
- Antigen Symbol:
- SF3B6
- SF3B6
- Antigen Synonyms:
- Ht006|P14|PM14_HUMAN|Pre mRNA branch site protein p14|Pre-mRNA branch site protein p14|SAP14|SF3B 14 kDa subunit|SF3B14|SF3B14a|Splicing factor 3B|14 kDa subunit
- Ht006|P14|PM14_HUMAN|Pre mRNA branch site protein p14|Pre-mRNA branch site protein p14|SAP14|SF3B 14 kDa subunit|SF3B14|SF3B14a|Splicing factor 3B|14 kDa subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3B4
- Q9Y3B4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Splicing factor 3B subunit 6 protein (1-125AA)
- Recombinant Human Splicing factor 3B subunit 6 protein (1-125AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice