Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-DEDD Rabbit Recombinant Polyclonal Antibody
Anti-DEDD Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-094
Supplier:  AFG Bioscience
New Product
undefined
Anti-DEDD Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-094
Supplier:  AFG Bioscience
Supplier Number:  A19192-50UL
New Product

Specifications

  • Antigen Symbol:
    DEDD
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 µl
  • Form:
    Liquid
  • Gene ID:
    O75618
  • Antigen Synonyms:
    CASP8IP1|Death effector domain containing|Death effector domain containing protein|Death effector domain containing testicular molecule|Death effector domain-containing protein|Death effector domain-containing testicular molecule|DEDD 1|DEDD_HUMAN|DEDD1|DEDPro 1|DEDPro1|DEFT|FLDED 1|FLDED-1|FLDED1|KE 05|KE05|OTTHUMP00000031822
  • Storage Buffer:
    PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • Storage Temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human Death effector domain-containing protein (1-318AA)
  • Purification:
    Antigen affinity purified
  • Packaging:
    Vial

Specifications

About this item

Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: DEDD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human