Anti-ATP5J Rabbit Recombinant polyclonal Antibody
Catalog # 77851-622
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP5J
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P18859
- Antigen Synonyms:ATP synthase|H+ transporting|mitochondrial F0 complex|subunit F6|ATP synthase-coupling factor 6|mitochondrial|ATP5|ATP5A|ATP5J_HUMAN|ATPase subunit F6|ATPM|CF6|F6
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human ATP synthase-coupling factor 6, mitochondrial protein (1-108AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200
Type:
Antigen: ATP5J
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat