Anti-DHDDS Rabbit Recombinant Polyclonal Antibody
77854-143
New Product
- Antigen Symbol:DHDDS
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q86SQ9
- Antigen Synonyms:HDSDehydrodolichyl diphosphate synthase complex subunit EC 2.5.1.87|Cis-isoprenyltransferase|CIT|Cis-IPTase|Cis-prenyltransferase subunit hCIT|Epididymis tissue protein Li 189m
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Dehydrodolichyl diphosphate synthase complex subunit DHDDS protein (1-333AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: DHDDS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human