Anti-DDX39A Rabbit Recombinant Polyclonal Antibody
77854-074
New Product
- Antigen Symbol:DDX39A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O00148
- Antigen Synonyms:DDX39A|DDX39|ATP-dependent RNA helicase DDX39A|EC 3.6.4.13|DEAD box protein 39|Nuclear RNA helicase URH49
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ATP-dependent RNA helicase DDX39A protein (178-427AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: DDX39A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human