Anti-ATP6V0D1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-635
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP6V0D1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P61421
- Antigen Synonyms:32 kDa accessory protein| ATP6D| ATP6DV| ATP6V0D1| ATPase H+ transporting lysosomal (vacuolar proton pump) member D| ATPase H+ transporting lysosomal 38kD V0 subunit d| ATPase H+ transporting lysosomal 38kDa V0 subunit d1| ATPase H+ transporting lysosomal V0 subunit d1| H(+) transporting two sector ATPase subunit D| p39| V ATPase 40 KDa accessory protein| V ATPase AC39 subunit| V ATPase subunit d 1| V ATPase subunit D| V-ATPase 40 kDa accessory protein| V-ATPase AC39 subunit| V-ATPase subunit d 1| V-type proton ATPase subunit d 1| VA0D1_HUMAN| Vacuolar ATP synthase subunit d 1| Vacuolar proton pump subunit d 1| VATX| VMA 6| VMA6| VPATPD
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human V-type proton ATPase subunit d 1 protein (1-351AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: ATP6V0D1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human