Anti-YBX1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, IHC:1:25 to 1:100
Type:
Antigen: YBX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77849-891
- 77849-890
- Antigen Symbol:
- YBX1
- YBX1
- Antigen Synonyms:
- BP 8|CBF-A|CCAAT binding transcription factor I subunit A|CCAAT-binding transcription factor I subunit A|CSDA2|CSDB|DBPB|DNA binding protein B|DNA-binding protein B|EFI-A|Enhancer factor I subunit A|MDR NF1|MGC104858|MGC110976|MGC117250|NSEP 1|NSEP1|Nuclease sensitive element binding protein 1|Nuclease-sensitive element-binding protein 1|p50|Q15905|Y-box binding protein 1|Y-box transcription factor|Y-box-binding protein 1|YB 1|YB-1|YBOX1_HUMAN|YBX 1|ybx1
- BP 8|CBF-A|CCAAT binding transcription factor I subunit A|CCAAT-binding transcription factor I subunit A|CSDA2|CSDB|DBPB|DNA binding protein B|DNA-binding protein B|EFI-A|Enhancer factor I subunit A|MDR NF1|MGC104858|MGC110976|MGC117250|NSEP 1|NSEP1|Nuclease sensitive element binding protein 1|Nuclease-sensitive element-binding protein 1|p50|Q15905|Y-box binding protein 1|Y-box transcription factor|Y-box-binding protein 1|YB 1|YB-1|YBOX1_HUMAN|YBX 1|ybx1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P67809
- P67809
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human YBX1
- Synthetic peptide of Human YBX1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice