Anti-WASF2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WASF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-524
- 77845-523
- Antigen Symbol:
- WASF2
- WASF2
- Antigen Synonyms:
- DICTYOSTELIUM|dJ393P12.2|IMD2|Protein WAVE-2|Putative Wiskott Aldrich syndrome protein family member 4|SCAR 2|SCAR|SCAR2|Suppressor of cyclic-AMP receptor (WASP family)|Verprolin homology domain-containing protein 2|WASF2_HUMAN|WASF4|WASP family protein member 2|WASP family protein member 4|WASP family Verprolin homologous protein 2|WASP-family protein member 2|WAVE 2|WAVE2|Wiskott-Aldrich syndrome protein family member 2|Wiskott-Aldrich syndrome protein family verprolin-homologous protein
- DICTYOSTELIUM|dJ393P12.2|IMD2|Protein WAVE-2|Putative Wiskott Aldrich syndrome protein family member 4|SCAR 2|SCAR|SCAR2|Suppressor of cyclic-AMP receptor (WASP family)|Verprolin homology domain-containing protein 2|WASF2_HUMAN|WASF4|WASP family protein member 2|WASP family protein member 4|WASP family Verprolin homologous protein 2|WASP-family protein member 2|WAVE 2|WAVE2|Wiskott-Aldrich syndrome protein family member 2|Wiskott-Aldrich syndrome protein family verprolin-homologous protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6W5
- Q9Y6W5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Wiskott-Aldrich syndrome protein family member 2 protein (243-317AA)
- Recombinant Human Wiskott-Aldrich syndrome protein family member 2 protein (243-317AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice