Anti-TRPC4AP Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: TRPC4AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77849-394
- 77849-395
- Antigen Symbol:
- TRPC4AP
- TRPC4AP
- Antigen Synonyms:
- antibody| Chromosome 20 open reading frame 188| Protein TAP1| Protein TRUSS| Short transient receptor potential channel 4 associated protein| Short transient receptor potential channel 4-associated protein| TAP1 protein| TNF receptor ubiquitous scaffolding/signaling protein| TNF-receptor ubiquitous scaffolding/signaling protein| TP4AP_HUMAN| Transient receptor potential cation channel| subfamily C| member 4 associated protein| Trp4 associated protein| Trp4-associated protein| Trpc4 associated protein| Trpc4-associated protein| TRRP4AP| TRUSS| Tumor necrosis factor receptor-associated ubiquitous scaffolding and signaling protein
- antibody| Chromosome 20 open reading frame 188| Protein TAP1| Protein TRUSS| Short transient receptor potential channel 4 associated protein| Short transient receptor potential channel 4-associated protein| TAP1 protein| TNF receptor ubiquitous scaffolding/signaling protein| TNF-receptor ubiquitous scaffolding/signaling protein| TP4AP_HUMAN| Transient receptor potential cation channel| subfamily C| member 4 associated protein| Trp4 associated protein| Trp4-associated protein| Trpc4 associated protein| Trpc4-associated protein| TRRP4AP| TRUSS| Tumor necrosis factor receptor-associated ubiquitous scaffolding and signaling protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TEL6
- Q8TEL6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TRPC4AP
- Fusion protein of Human TRPC4AP
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice