Anti-TRHDE Rabbit Recombinant polyclonal Antibody
Catalog # 77843-892
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TRHDE
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9UKU6
- Antigen Synonyms:UNQ2507/PRO5995|Thyrotropin-releasing hormone-degrading ectoenzyme|TRH-DE|TRH-degrading ectoenzyme|EC 3.4.19.6|Pyroglutamyl-peptidase II|PAP-II|TRH-specific aminopeptidase|Thyroliberinase
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Thyrotropin-releasing hormone-degrading ectoenzyme protein (62-200AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: TRHDE
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat