Ascorbate Oxidase (from Cucurbita sp.)
Catalog # 80050-362
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:9029-44-1
- Source:Cucurbita sp.
- Size:1000 U
- Storage Conditions:Desiccated (hygroscopic), OK to freeze
- Enzyme Name:Ascorbate Oxidase
- Enzyme Synonyms:Ascorbate Oxidase, Cucurbita sp.