Anti-COL12A1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77853-372
New Product
- Antigen Symbol:COL12A1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Chicken
- Form:Liquid
- Gene ID:P13944
- Antigen Synonyms:COL12A1|Collagen alpha-1(XII) chain|Fibrochimerin
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Chicken Collagen alpha-1(XII) chain protein (139-311AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Type:
Antigen: COL12A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken