Anti-GLIPR1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:100 to 1:300
Type:
Antigen: GLIPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77848-909
- 77848-908
- Antigen Symbol:
- GLIPR1
- GLIPR1
- Antigen Synonyms:
- CRISP 7|CRISP7|GLI pathogenesis related 1|GLI pathogenesis-related 1 (glioma)|Glioma pathogenesis related protein 1|Glioma pathogenesis-related protein 1|GLIP1_HUMAN|GliPR 1|GLIPR|Protein RTVP-1|RELATED TO TESTIS SPECIFIC|VESPID|AND PATHOGENESIS PROTEINS 1|RTVP 1|RTVP1|Testes specific vespid and pathogenesis protein 1|testes-specific vespid and pathogenesis protein 1
- CRISP 7|CRISP7|GLI pathogenesis related 1|GLI pathogenesis-related 1 (glioma)|Glioma pathogenesis related protein 1|Glioma pathogenesis-related protein 1|GLIP1_HUMAN|GliPR 1|GLIPR|Protein RTVP-1|RELATED TO TESTIS SPECIFIC|VESPID|AND PATHOGENESIS PROTEINS 1|RTVP 1|RTVP1|Testes specific vespid and pathogenesis protein 1|testes-specific vespid and pathogenesis protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48060
- P48060
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human GLIPR1
- Fusion protein of Human GLIPR1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice