Anti-INTU Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: INTU
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-280
- 77844-281
- Antigen Symbol:
- INTU
- INTU
- Antigen Synonyms:
- Homolog of inturned (X. tropicalis)|INT|Inturned planar cell polarity effector homolog|KIAA1284|PDZ domain containing 6|PDZ domain-containing protein 6|PDZD 6|PDZD6_HUMAN|PDZK6
- Homolog of inturned (X. tropicalis)|INT|Inturned planar cell polarity effector homolog|KIAA1284|PDZ domain containing 6|PDZ domain-containing protein 6|PDZD 6|PDZD6_HUMAN|PDZK6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULD6
- Q9ULD6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein inturned protein (6-139AA)
- Recombinant Human Protein inturned protein (6-139AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice