Rat Recombinant Geranylgeranyltransferase I (from E. coli)
80603-168
:
- Pack type:plastic ampoule
- Enzyme type:Recombinant
- Source:E. coli
- Species:Rat
- Size:50 μg
- Storage Conditions:Safe to freeze, avoid repeated freeze/thaw cycles.
- Enzyme Name:Geranylgeranyltransferase I
- Enzyme Synonyms:GGTase I
- Purity:≥90% (SDS-PAGE)